D-(+)-Sucrose, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

Management of Change (MOC) category = N

J.T. Baker®
BAKR4072-07 BAKR4072-05 4072-09 4072-01 BAKR4072-01 4072-05 BAKR4072-09 4072-07
CAJT4072-09CS 2278.72 CAD
JT4072-09 CAJT4072-09 CAJT4072-5 JT4072-5 CAJT4072-1 JT4072-7 JT4072-1 CAJT4072-7
D-(+)-Sucrose, crystals, BAKER ANALYZED® ACS, J.T.Baker®
D(+)-Sucrose

Protect and stabilize valuable biochemical proteins and streamline your downstream processing with reliable, high-performance sugars.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00006626
CAS Number: 57-50-1
Merck Index: 13,08966
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Specific Rotation (+) 66.3 - 66.8 Degree
ACS - Insoluble Matter ≤0.005 %
ACS - Loss on Drying at 105°C ≤0.03 %
ACS - Residue after Ignition ≤0.01 %
ACS - Titrable Acid (meq/g) ≤0.0008
ACS - Chloride (Cl) ≤0.005 %
ACS - Sulfate and Sulfite (as SO₄) ≤0.005 %
ACS - Invert Sugar ≤0.05 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR