Sodium chloride ≥99%, crystals, Ultrapure Bioreagent for biochemistry, for chromatography, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
BAKR4058-01
4058-02
CAJT4058-1EA
148.54
CAD
CAJT4058-1
CAJT4058-5
CAJT4058-7
JT4058-02
CAJT4058-6
Sodium chloride ≥99%, crystals, Ultrapure Bioreagent for biochemistry, for chromatography, J.T.Baker®
Sodium chloride
ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.
Formula:
NaCl Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (NaCl) | ≥99.0 % |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Protease Activity | Passes Test |
pH of 5% Solution at 25°C | 5.5 - 7.5 |
Insoluble Matter | ≤0.005 % |
Bromide (Br) | ≤0.01 % |
Chlorate and Nitrate (as NO₃) | ≤0.003 % |
Iodide (I) | ≤0.002 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
Barium (Ba) | ≤0.001 % |
Calcium, Magnesium, and R₂O₃ Precipitate | ≤0.005 % |
Potassium (K) | ≤0.005 % |
Nitrogen Compounds (as N) | ≤5 ppm |
Phosphate (PO₄) | ≤5 ppm |
Heavy Metals (as Pb) | ≤2 ppm |
Trace Impurities - Iron (Fe) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...