Strontium chloride hexahydrate 99.0-103.0%, crystals, BAKER ANALYZED® ACS low in magnesium

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR4036-04 BAKR4036-01 BAKR4036-05
CAJT4036-4EA 1680.14 CAD
CAJT4036-4 CAJT4036-1 CAJT4036-5
Strontium chloride hexahydrate 99.0-103.0%, crystals, BAKER ANALYZED® ACS low in magnesium
Strontium chloride hexahydrate

Crystals. Low in magnesium.

Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
Boiling Pt: 100
Melting Pt: 61
Density: 1.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (SrCl₂·6H₂O) 99.0 - 103.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 7.0
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Barium (Ba) ≤0.05 %
ACS - Calcium (Ca) ≤0.05 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Magnesium (Mg) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR