Sodium dihydrogen phosphate monohydrate 99, crystals, ultrapure_bioreagent LC, LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

M.O.C. Category NI

BAKR4011-01 BAKR4011-05
CAJT4011-1EA 385.2 CAD
CAJT4011-1 CAJT4011-5
Sodium dihydrogen phosphate monohydrate 99, crystals, ultrapure_bioreagent LC, LC-MS, J.T.Baker®
Sodium dihydrogen phosphate monohydrate

For liquid chromatography and molecular biology applications. Lot analysis on label.

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaH₂PO₄·H₂O) 99.0 - 102.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 4.1 - 4.5
Insoluble Matter, Calcium and NH₄OH Precipitate ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR