Trisodium phosphate dodecahydrate 98.0-102.0% (by acidimetry), crystals, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

BAKR3836-07 BAKR3836-01 3836-07
CAJT3836-7EA 2967.45 CAD
CAJT3836-7 CAJT3836-1 CAJT3836-5 JT3836-7
Trisodium phosphate dodecahydrate 98.0-102.0% (by acidimetry), crystals, BAKER ANALYZED® ACS
tri-Sodium phosphate dodecahydrate

Crystals. Lot analysis on label.

Formula: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: ∼75 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
UN: 3262
ADR: 8 III
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₃PO₄·12H₂O) (by acidimetry) 98.0 - 102.0 %
Excess Alkali (as NaOH) ≤2.5 %
Insoluble Matter ≤0.01 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR