KIMAX® Kohlrausch Volumetric Flasks, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28100-100 28100-200 28100-500
89002-068CS 2624.87 CAD
89002-068 89002-070 89002-072
KIMAX® Kohlrausch Volumetric Flasks, Class A, Kimble Chase
Flasks Volumetric Flasks
These flasks feature an enlarged mouth to ease the introduction of solid material samples
Graduation ring is blasted on the neck. Flasks are calibrated to contain and have a marking spot.
Order Now
SPECIFICATIONS
Environmentally Preferable