Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC endotoxin tested, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium dihydrogen orthophosphate monohydrateSodium dihydrogenorthophosphate monohydrateMonosodium phosphate monohydrateSodium phosphate monobasic monohydrateSodium orthophosphate monobasic monohydrate

Management of Change (MOC) category = HR

BAKR3821-07 BAKR3821-09 3821-QM 3821-09 BAKR3821-01 3821-01 BAKR3821-QM 3821-07
CAJT3821-7EA 1631.54 CAD
JT3821-1 JT3821-QM JT3821-09 CAJT3821-7 CAJT3821-09 CAJT3821-QM JT3821-7 CAJT3821-1
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC endotoxin tested, J.T.Baker®
Sodium dihydrogen phosphate monohydrate

  • Food GMP manufactured product

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149208
CAS Number: 10049-21-5
REACH: 01-2119489796-13
Merck Index: 13,08734
https://ca.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
FCC - Assay (anhydrous basis) 98.0 - 103.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Fluoride (F) ≤0.005 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 10.0 - 15.0 %
Endotoxin Concentration (EU/g) ≤2.5
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR