Sodium dihydrogen phosphate monohydrate ≥98% (by anhydrous basis), crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

Management of Change (MOC) category = HR

BAKR3820-07 3820-01 BAKR3820-09
CAJT3820-7EA 4855.51 CAD
CAJT3820-7 CAJT3820-1 JT3820-1 CAJT3820-09
Sodium dihydrogen phosphate monohydrate ≥98% (by anhydrous basis), crystals USP, FCC, J.T.Baker®
Sodium dihydrogen phosphate monohydrate

Crystals. GMP manufactured.


  • Food GMP manufactured product

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
FCC - Assay (anhydrous basis) 98.0 - 103.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Fluoride (F) ≤0.005 %
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 10.0 - 15.0 %
Appearance (White crystals or granules) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR