Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
Management of Change (MOC) category = NI
BAKR3819-01
BAKR3819-05
CAJT3819-1EA
361.58
CAD
CAJT3819-1
CAJT3819-5
Sodium dihydrogen phosphate dihydrate 98.0-102.0%, BAKER ANALYZED®
Sodium dihydrogen phosphate dihydrate
Lot analysis on label.
Formula:
NaH₂PO₄·2H₂O Boiling Pt: ? 650 °C (1013 hPa) Melting Pt: 60 °C Density: 1.92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149209 CAS Number: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (NaH₂PO₄·2H₂O) | 98.0 - 102.0 % |
pH of 5% Solution at 25°C | 4.1 - 4.5 |
Chloride (Cl) | ≤5 ppm |
Insoluble Matter, Calcium and NH₄OH Precipitate | ≤0.01 % |
Nitrogen Compounds (as N) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
Arsenic (As) | ≤0.5 ppm |
Heavy Metals (as Pb) | ≤0.001 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...