Viton® Unsupported Gloves, Showa
Supplier: Showa-Best Glove
892-07 890-09 890-09
28888-330PK 223.71 CAD
28888-330 28888-332 CA28888-328 CA32892-048 CA32892-049
Viton® Unsupported Gloves, Showa
Gloves
Best® Viton® Fluoroelastomer rubber provides superior resistance to highly corrosive acids.
  • Excellent for handling aliphatic and aromatic hydrocarbons
  • Chemical resistant glove
This synthetic rubber provides the highest permeation resistance to high molecular aromatic hydrocarbons such as Benzene, Toluene, Xylene and Polychlorinated Biphenyls (PCBs) of any protective material used to make gloves.
These are used in military applications, acids and caustics, chlorinated solvents fuels etc.
Viton gloves provide the ultimate combination of protection and dexterity.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable