PYREX® Volumetric Flasks, Class A, Micro, [ST] Stopper, Individually Serialized, Corning
Supplier: Corning
5631-1 5631-10 5631-2 5631-25
77777-070CS 980.75 CAD
77777-070 77777-072 77777-074 77777-076
PYREX® Volumetric Flasks, Class A, Micro, [ST] Stopper, Individually Serialized, Corning
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with glass stoppers.
  • Hexagonal base to improve stability
  • Sharp and permanent markings
  • White markings for easy reading
  • With certificate of identification and capacity
  • Ideal for microchemical works
Certifications: ASTM E-237