Sodium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR3770-09 BAKR3770-07 BAKR3770-01 BAKR3770-05
CAJT3770-09EA 5778.94 CAD
CAJT3770-09 CAJT3770-7 CAJT3770-1 CAJT3770-5
Sodium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium nitrate

Crystals. Lot analysis on label.

Formula: NaNO₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NaNO₃) ≥99.0 %
ACS - pH of 5% Solution at 25°C 5.5 - 8.3
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
ACS - Iodate (IO₃) ≤5 ppm
ACS- Nitrite (NO₂) ≤0.001 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sulfate (SO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤3 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR