Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Sodium cobaltnitrite, Trisodium hexanitritocobaltate
Management of Change (MOC) category = N
BAKR3656-01
3656-04
CAJT3656-1EA
1078.63
CAD
CAJT3656-1
JT3656-4
Sodium hexanitritocobaltate(III), BAKER ANALYZED® ACS, J.T.Baker®
Sodium hexanitritocobaltate(III)
For determination of potassium.
- Lot analysis on label
Formula:
Na₃Co(NO₂)₆ MW: 403.94 g/mol Melting Pt: ∼220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003512 CAS Number: 13600-98-1 EINECS: 237-077-7 UN: 1479 ADR: 5.1 II |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For R&D use only. Not on TSCA Inventory. | |
Insoluble Matter | ≤0.02 % |
Suitability for Potassium Determination | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...