tri-Sodium citrate dihydrate 99 Anhydrous basis, granules FCC, USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

M.O.C. Category R

BAKR3649-07 BAKR3649-08 BAKR3649-05 3649-01
CAJT3649-7EA 2582.35 CAD
CAJT3649-7 CAJT3649-8 CAJT3649-1 CAJT3649-5 JT3649-1
tri-Sodium citrate dihydrate 99 Anhydrous basis, granules FCC, USP, J.T.Baker®
tri-Sodium citrate dihydrate

Granular. GMP manufactured.


  • GMP manufactured product


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Certifications: Meets USP and FCC requirements.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.70 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Alkalinity Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Tartrate Passes Test
FCC - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Alkalinity Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Water (FCC) 10.0 - 13.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR