KIMAX® Volumetric Flasks with [ST] Glass Stopper, Red Stripe, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28013-100 28013-1000 28013-200 28013-2000 28013-25 28013-250 28013-50 28013-500
89001-898CS 1378.61 CAD
89001-898 89001-900 89001-902 89001-904 89001-906 89001-908 89001-910 89001-912
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Red Stripe, Class A, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288, Class A unserialized requirements.
Graduation line is etched through a vertical red stripe on neck. Contrast between graduation line and deep amber background provides legibility without coloring the graduation line. Supplied with [ST] ground glass stopper.