tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granules USP, JP, Ph. Eur., BP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

Management of Change (MOC) category = R

BAKR3647-20 BAKR3647-07 BAKR3647-05
CAJT3647-20EA 5176.82 CAD
CAJT3647-20 CAJT3647-7 CAJT3647-1 CAJT3647-5
tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granules USP, JP, Ph. Eur., BP, Multi-Compendial, J.T.Baker®
tri-Sodium citrate dihydrate

Granular. GMP manufactured.


  • GMP manufactured product

Formula: HOC(COONa)(CH₂COONa)₂·2H₂O
MW: 294.1 g/mol
Melting Pt: 150
Density: 1.70 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Alkalinity Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Tartrate Passes Test
USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Chloride (Cl) ≤50 ppm
EP/BP - Oxalate (C₂O₄) ≤300 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Water (H₂O) 11.0 - 13.0 %
EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
EP/BP - Endotoxin Concentration (EU/g) ≤2.5
JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
JP - pH (1 in 20) 7.5 - 8.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.015 %
JP - Identification Passes Test
JP - Sulfate (SO₄) ≤0.048 %
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Tartrate Passes Test
JP - Oxalate (C₂O₄) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Loss on Drying at 180°C 10.0 - 13.0 %
Total Aerobic Microbial Count, For Information Only
Total Yeasts and Molds Count, For Information Only
E. coli Negative
P. aureginosa Negative
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR