KIMAX® Volumetric Flasks with Snap Cap, Class B, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28010-10 28010-100 28010-1000 28010-200 28010-25 28010-50 28010-500
89001-870CS 682.2 CAD
89001-870 89001-872 89001-874 89001-876 89001-878 89001-880 89001-882 89001-884 89001-886
KIMAX® Volumetric Flasks with Snap Cap, Class B, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288, Class B requirements.
Flasks have a graduation ring blasted on the neck and a marking spot. Neck opening is tooled for the supplied polyethylene snap cap. The enlarged top of cap protects the neck if flask is tipped over.