Sodium chloride 99-100.5% (dried basis), granular USP, FCC endotoxin tested, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Saline, NaCl, Sea salt, Halite
Management of Change (MOC) category = R
BAKR3629-07
BAKR3629-09
CAJT3629-7EA
819.93
CAD
CAJT3629-7
CAJT3629-9
Sodium chloride 99-100.5% (dried basis), granular USP, FCC endotoxin tested, J.T.Baker®
Sodium chloride
Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the products are cGMP produced and meet USP grade standards.
- Food GMP manufactured product
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
USP - Assay (NaCl) (USP) (dried basis) | 99.0 - 100.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Acidity or Alkalinity | Passes Test |
USP - Aluminum (Al) | ≤0.2 ppm |
USP - Arsenic (As) | ≤1 ppm |
USP - Appearance of Solution | Passes Test |
USP - Barium (Ba) | Passes Test |
USP - Endotoxin Concentration (2.5 EU/g max.) | Passes Test |
USP - Nitrite (NO₂) (Absorbance Value) | ≤0.01 |
USP - Iron (Fe) | ≤2 ppm |
USP - Magnesium and Alkaline Earth Metals (as Ca) | ≤100 ppm |
USP - Loss on Drying | ≤0.5 % |
USP - Iodide (I) | Passes Test |
USP - Bromide (Br) | ≤100 ppm |
USP - Ferrocyanide | Passes Test |
USP - Phosphate (PO₄) | ≤25 ppm |
USP - Potassium (K) | ≤500 ppm |
USP - Sulfate (SO₄) | ≤200 ppm |
FCC - Assay (as NaCl) (FCC) (ignited basis) | 99.0 - 100.5 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Arsenic (As) | ≤1 ppm |
FCC - Calcium and Magnesium (as Ca) (by EDTA titration)(FCC) | ≤0.35 % |
FCC - Heavy Metals (as Pb) | ≤2 ppm |
FCC - Iron (Fe) | ≤2 ppm |
FCC - Loss on Drying | ≤0.5 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Must be subjected to further processing during the preparation of | |
injectable dosage forms | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...