Sodium carbonate, anhydrous ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR3604-07 BAKR3604-05 3604-01
CAJT3604-7EA 1433.23 CAD
CAJT3604-7 CAJT3604-1 CAJT3604-5 JT3604-1
Sodium carbonate, anhydrous ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium carbonate

Granular. Lot analysis on label.

Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂CO₃) (dried basis)(by acidimetry) ≥99.5 %
Insoluble Matter ≤0.01 %
Loss on Heating at 285°C ≤1.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Silica (SiO₂) ≤0.005 %
Sulfur Compounds (as SO₄) ≤0.003 %
Calcium (Ca) ≤0.03 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR