Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals FCC, NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium carbonate hydrousSoda monohydrate

Management of Change (MOC) category = R

3600-05 BAKR3600-S1 3600-09 BAKR3600-01 3600-D1 BAKR3600-09 BAKR3600-QM BAKR3600-S1 BAKR3600-05 3600-QM 3600-S1 3600-D1 3600-01
CAJT3600-5EA 821.25 CAD
JT3600-9 CAJT3600-5 JT3600-D1 CAJT3600-9 CAJT3600-D1 JT3600-1 CAJT3600-S1 JT3600-5 TXLJT3600S1CTT JT3600-QM JT3600-S1 CAJT3600-QM CAJT3600-1
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals FCC, NF, J.T.Baker®
Sodium carbonate monohydrate
Formula: Na₂CO₃·1H₂O
MW: 124.00 g/mol
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
MDL Number: MFCD00149177
CAS Number: 5968-11-6

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) 12.0 - 15.0 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 12.0 - 15.0 %
FCC - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR