Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
BAKR3588-07
BAKR3588-01
3588-05
CAJT3588-7EA
2104.35
CAD
CAJT3588-7
CAJT3588-1
CAJT3588-5
JT3588-5
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS
Sodium bromide
Crystals. Lot analysis on label.
- Lot analysis on label
Formula:
NaBr Boiling Pt: 1396 °C (1013 hPa) Melting Pt: 753 °C Density: 3.21 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 Merck Index: 13,08666 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (NaBr) | ≥99.0 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Alkalinity (as Na₂CO₃) | ≤0.015 % |
ACS - pH of 5% Solution at 25°C | 5.0 - 8.8 |
ACS - Bromate (BrO₃) | ≤0.001 % |
ACS - Calcium (Ca) | ≤0.002 % |
ACS - Chloride (Cl) | ≤0.2 % |
ACS - Sulfate (SO₄) | ≤0.002 % |
ACS - Barium (Ba) | ≤0.002 % |
ACS - Magnesium (Mg) | ≤0.001 % |
ACS - Potassium (K) | ≤0.1 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...