di-Sodium tetraborate decahydrate ≥99% NF
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
Management of Change (MOC) category = R
BAKR3574-07
BAKR3574-05
3574-01
CAJT3574-7EA
1583.79
CAD
CAJT3574-7
CAJT3574-1
CAJT3574-5
JT3574-1
di-Sodium tetraborate decahydrate ≥99% NF
di-Sodium tetraborate decahydrate
Powder.
- GMP manufactured product
Formula:
Na₂B₄O₇·10H₂O MW: 381.37 g/mol Boiling Pt: 320 Melting Pt: 75 Density: 1.73 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 13,08662 |
Specification Test Results
Meets N.F. Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay | 99.0 - 105.0 % |
NF - Identification | Passes Test |
NF - Carbonate and Bicarbonate | Passes Test |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...