Sodium metabisulfite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

Management of Change (MOC) category = N

BAKR3552-01 BAKR3552-05 BAKR3552-07 3552-01
CAJT3552-1CS 2899.49 CAD
CAJT3552-1 CAJT3552-5 CAJT3552-7 JT3552-1
Sodium metabisulfite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Sodium metabisulphite

Granular. Lot analysis on label.

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
Merck Index: 13,08712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂S₂O₅) ≥97.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.05 %
Thiosulfate (S₂O₃) ≤0.05 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR