Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

BAKR3510-07 BAKR3510-09 3510-05
CAJT3510-7EA 1079.33 CAD
CAJT3510-7 CAJT3510-9 CAJT3510-5 JT3510-5
Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
Sodium hydrogen carbonate

Powder. Pyrogen tested. GMP manufactured.

Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification Passes Test
USP - Limit of Sulfur Compounds ≤ 0.015 %
USP - Loss on Drying ≤ 0.25 %
USP - Insoluble Matter Passes Test
USP - Normal Carbonate Passes Test
USP - Limit of Ammonia ≤ 20 ppm
USP - Chloride (Cl) ≤ 0.015 %
USP - Arsenic (As) ≤ 2 ppm
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Assay (NaHCO₃) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Carbonate (CO₃) Passes Test
EP/BP - Chloride (Cl) ≤ 150 ppm
EP/BP - Sulfate (SO₄) ≤ 150 ppm
EP/BP - Ammonium (NH₄) ≤ 20 ppm
EP/BP - Calcium (Ca) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 20 ppm
JP - Assay (NaHCO₃) 99.0 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 7.9 - 8.4
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.040 %
JP - Carbonate (CO₃) Passes Test
JP - Ammonium (NH₄) Passes Test
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Arsenic (As) ≤ 2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR