Sodium hydrogen carbonate ≥99% (dried basis), powder USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

BAKR3509-01 BAKR3509-09 BAKR3509-05 3509-07
CAJT3509-1EA 130.52 CAD
CAJT3509-1 CAJT3509-7 CAJT3509-9 CAJT3509-5 JT3509-7
Sodium hydrogen carbonate ≥99% (dried basis), powder USP, FCC, J.T.Baker®
Sodium hydrogen carbonate

Powder. GMP manufactured. Not for use in hemodialysis.


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Certifications: Meets USP and FCC requirements.


Caution: Not for use in hemodialysis.

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Density: 2.20 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Identification Passes Test
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Insoluble Substances Passes Test
USP - Normal Carbonate Passes Test
USP - Limit of Ammonia ≤20 ppm
USP - Chloride (Cl) ≤0.015 %
USP - Arsenic (As) ≤2 ppm
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying ≤0.25 %
FCC - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
Appearance (White powder) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is not intended for use in hemodialysis.
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR