Sodium benzoate 99.0-101.0% (by anhydrous basis) FCC, NF, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Benzoic acid sodium salt
Management of Change (MOC) category = R
BAKR3500-01
3500-05
BAKR3500-05
3500-01
CAJT3500-5EA
930.08
CAD
CAJT3500-5
JT3500-1
CAJT3500-1
JT3500-5
Sodium benzoate 99.0-101.0% (by anhydrous basis) FCC, NF, J.T.Baker®
Sodium benzoate
Formula:
C₆H₅COONa MW: 144.11 g/mol Melting Pt: 410…430 °C Density: 1.44 g/cm³ (20 °C) Flash Pt: >100 °C |
MDL Number:
MFCD00012463 CAS Number: 532-32-1 Merck Index: 13,08654 |
Specification Test Results
Meets N.F. Requirements | |
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Identification C | Passes Test |
NF - Alkalinity | Passes Test |
NF - Water (H₂O) | ≤1.5 % |
NF - Assay (C₆H₅COONa) (anhydrous basis) | 99.0 - 101.0 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Identification C | Passes Test |
FCC - Alkalinity | ≤0.04 % |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Water (H₂O) | ≤1.5 % |
FCC - Assay (C₆H₅COONa) (anhydrous basis) | 99.0 - 101.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...