Sodium acetate, anhydrous ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = NI
BAKR3470-07
BAKR3470-R
3470-05
3470-01
CAJT3470-7EA
2664.05
CAD
CAJT3470-7
CAJT3470-R
CAJT3470-1
CAJT3470-5
JT3470-5
JT3470-1
Sodium acetate, anhydrous ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium acetate
Lot analysis on label.
- Lot analysis on label
Formula:
H₃CCOONa Boiling Pt: 881.4 °C (1013 hPa) Melting Pt: 324 °C Density: 1.53 g/cm³ Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 REACH: 01-2119485123-42 Merck Index: 12,08711 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃COONa) | ≥99.0 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Loss on Drying at 120°C | ≤1.0 % |
ACS - pH of 5% Solution at 25°C | 7.0 - 9.2 |
ACS - Chloride (Cl) | ≤0.002 % |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
ACS - Calcium (Ca) | ≤0.005 % |
ACS - Heavy Metals (as Pb) | ≤0.001 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - Magnesium (Mg) | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...