Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR3401-01 BAKR3401-05 3401-05
CAJT3401-5EA 1170.7 CAD
CAJT3401-5 JT3401-5 JT3401-1
Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®
Silica gel

6 to 18 mesh. Indicating (cobalt chloride). Lot analysis on label.


Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.10 kg/m³
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Loss on Heating at 200°C ≤6.0 %
Suitability for Moisture Absorption ≥27 %
Suitability as Indicating Desiccant Passes Test
Mesh - Thru U.S. No. 6 Sieve ≥90 %
Mesh - Thru U.S. No. 18 Sieve ≤10 %
Color Change: Blue.................Activated Violet..........10% Moisture Absorbed Pink............19% Moisture Absorbed Pale Pink.......28% Moisture Absorbed

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR