Sodium sulfate, anhydrous 99, granules, ULTRA RESI-ANALYZED™, ACS Reagent ACS 12-60 Mesh Pest_Residual, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

M.O.C. Category N

BAKR3375-07 BAKR3375-09 BAKR3375-05 3375-09 3375-01
CAJT3375-7EA 1650.01 CAD
CAJT3375-7 CAJT3375-9 CAJT3375-1 CAJT3375-5 JT3375-9 JT3375-1
Sodium sulfate, anhydrous 99, granules, ULTRA RESI-ANALYZED™, ACS Reagent ACS 12-60 Mesh Pest_Residual, J.T.Baker®
Sodium sulphate

BAKER ULTRA RESI-ANALYZED® Reagent ultra high purity solvents and salts developed for organic residue extraction/concentration procedures.

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂SO₄) ≥99.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Ignition ≤0.5 %
ACS - pH of 5% Solution at 25°C 5.2 - 9.2
ACS - Chloride (Cl) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Calcium (Ca) ≤0.01 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Extraction-Concentration Suitability * Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Nitrogen Compounds (as N) ≤5 ppm
* Result obtained on a sample dried at 400°C for one hour (ACS method)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR