5-Sulfosalicylic acid dihydrate 99.0-101.0%, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR0364-01 BAKR0364-05
CAJT0364-1CS 2473.35 CAD
CAJT0364-1 CAJT0364-5
5-Sulfosalicylic acid dihydrate 99.0-101.0%, BAKER ANALYZED® ACS
5-Sulphosalicylic acid dihydrate

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Melting Pt: 120 °C
Density: 1.695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=6a3840cf38254959a0062041e9a84fa6740b1288&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₇H₆O₆S·2H₂O) 99.0 - 101.0 %
Insoluble Matter ≤0.02 %
Residue after Ignition ≤0.10 %
Chloride (Cl) ≤0.001 %
Salicylic Acid (HOC₆H₄COOH) ≤0.04 %
Sulfate (SO₄) ≤0.02 %
Heavy Metals (as Pb) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
STORAGE: Keep product from light.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR