Sodium hydroxide 25% in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR0338-09 BAKR0338-07 BAKR0338-26
CA0338-9EA 17110.1 CAD
CA0338-9 CAJT0338-07 CAJT0338-26
Sodium hydroxide 25% in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from USP purified water and multicompendial sodium hydroxide pellets. Filtered through a 0.2 µm element. GMP manufactured product.

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NaOH) 24.5 - 25.5 %
Chloride (Cl) ≤20 ppm
Heavy Metals (as Pb) ≤1 ppm
Iron (Fe) ≤2 ppm
Endotoxin Concentration (EU/mL) ≤2.0
Made from USP Purified Water and Sodium Hydroxide pellets, NF (Multicompendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR