D-(+)-Sucrose, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®

Supplier: MilliporeSigma

Synonyms: α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranosideSucroseD(+)-Saccharoseβ-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranosideα-ᴅ-Glc-(1→2)-β-ᴅ-Fru

1.07653.9013 1.07653.9029 1.07653.9053 1.07653.9029 1.07653.9013 1.07653.9525 1.07653.5000
CA1.07653.9013EA 752.76 CAD
EM1.07653.9029 CA1.07653.9013 CA1.07653.5000 EM1.07653.9013 CA1.07653.9029 CA1.07653.9525 EM1.07653.9053
D-(+)-Sucrose, EMPROVE® ESSENTIAL BP, JP, NF, Ph. Eur., SAFC®
D(+)-Sucrose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00006626
CAS Number: 57-50-1
Merck Index: 13,08966
https://ca.vwr.com/stibo/web/std.lang.all/95/86/2299586.jpg

Order Now

Specification Test Results

Acidity or alkalinity Passes test
Appearance of solution (50%, water) Passes test
Arsenic (As) 0.0001% max
Calcium (Ca) Passes
Chloride (Cl) 0.0035% max
Color number (Ph. Eur.) 45 max
Dextrins Passes test
Electrical conductivity 35µS/cm max
Endotoxins 250I.U./g max
Heavy metals (as Pb) 0.0005% max
Identity Passes
Lead (Pb) 0.00005% max
Loss on drying (105°C) 0.1% max
Organic volatile impurities (according to NF) Conforms
Other residual solvents (Ph.Eur/USP/ICH) Excluded by manufacturing process
Reducing sugars/invert sugar Passes
Residual solvents (Ph.Eur/USP/ICH) of class 3 0.5% max
Specific rotation [a]20/D (26%, water) +66.3-+67.0°
Sulfate (SO4) 0.005% max
Sulfated ash 0.02% max
Sulfite (SO2) 0.0010% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR