Chloroflex™ Flock-Lined Neoprene Gloves, Showa
Supplier: Avantor
723M-08 723XL-10
32892-054CS 2133.6 CAD
32892-054 32892-055 CA32892-053 CA32892-054 CA32892-055 CA32892-056
Chloroflex™ Flock-Lined Neoprene Gloves, Showa
Gloves
Premium quality neoprene is case-hardened and chemical-resistant, resisting effects from a wide range of oils and chemicals, heat, sunlight, ozone, and aging.
  • Protects against abrasion, cuts, and punctures
  • Rayon flock lining absorbs perspiration and allows easy donning and doffing
  • Provide hand protection from many acids, caustics, and most organic solvents
  • Tractor-tread grip
Color: black.
Certifications: In compliance with FDA 21 CFR 170-199.