AlphaTec® Butyl 38-514 Chemical resistant gloves, Ansell
Supplier: Ansell Canada
103201 103202 103203 103204
89011-854CS 8549.17 CAD
89011-854 89011-856 89011-858 89011-860
AlphaTec® Butyl 38-514 Chemical resistant gloves, Ansell
These gloves deliver superior resistance to esters, ketones, and oxidizing agents, including concentrated oxidizing mineral acids such as 70% nitric acid and 98% sulfuric acid.
  • Hand specific
  • Beaded cuffs
  • Superior resistance to chemicals
Ideal for hazmat, first response, unknown exposure, sampling, production, and maintenance applications. Each pair is packed with an insert containing critical information on appropriate usage.