Sodium hydroxide 99-100 alkilinity_total_NaOH, pellets, EMSURE® ACS, Reag. Ph Eur Reag. Ph. Eur., ACS LC-MS K_0,02, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda

MQ300

1.06469.9050 1.06469.9012 1.06469.1000 1.06469.5000
CA1.06469.9050EA 3003 CAD
CA1.06469.9050 CASX0590-7 CA1.06469.9012 CA1.06469.1000 CA1.06469.5000
Sodium hydroxide 99-100 alkilinity_total_NaOH, pellets, EMSURE® ACS, Reag. Ph Eur Reag. Ph. Eur., ACS LC-MS K_0,02, Supelco®
Sodium hydroxide

Pellets.


In North America 1.06469.9050 replaces and is identical to SX0590-20

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323 (lit.)
Density: 2.13 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (acidimetric,NaOH) 99 - 100 %
Assay (total alkalinity calc. as NaOH) 99 - 100 %
Identity passes test
Appearance of solution passes test
Carbonate (as Na₂CO₃) ≤ 0.5 %
Chloride (Cl) ≤ 0.0005 %
Phosphate (PO₄) ≤ 0.0005 %
Silicate (SiO₂) ≤ 0.001 %
Sulfate (SO₄) ≤ 0.0005 %
Total nitrogen (N) ≤ 0.0003 %
Heavy metals (as Ag) ≤ 0.0005 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Ca (Calcium) ≤ 0.0005 %
Cu (Copper) ≤ 0.0002 %
Fe (Iron) ≤ 0.0005 %
Hg (Mercury) ≤ 0.00001 %
K (Potassium) ≤ 0.020 %
Mg (Magnesium) ≤ 0.0005 %
Ni (Nickel) ≤ 0.0005 %
Pb (Lead) ≤ 0.0005 %
Zn (Zinc) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR