Sodium chloride 99,5-100,5 argentometric_dried, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
Synonyms:
Saline, NaCl, Sea salt, Halite
1.06400.9103
1.06400.5000
1.06400.9029
1.06400.1000
1.06400.9051
1.06400.9500
CA1.06400.9103EA
3480.33
CAD
CA1.06400.9103
CA1.06400.5000
CA1.06400.9029
CA1.06400.1000
CA1.06400.9051
CA1.06400.9500
Sodium chloride 99,5-100,5 argentometric_dried, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, SAFC®
Sodium chloride
Crystals. Soluble in water. For pharmaceutical production.
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Assay (argentometric; calculated on dried substance) | 99.5 - 100.5 % |
Identity | passes test |
Appearance of solution | passes test |
Acidity or alkalinity | passes test |
Bromide (Br) | ≤ 0.005 % |
Ferrocyanides | passes test |
Iodide (I) | passes test |
Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water) | ≤ 0.01 |
Phosphate (PO₄) | ≤ 0.0025 % |
Sulfate (SO₄) | ≤ 0.0200 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
As (Arsenic) | ≤ 0.0001 % |
Ba (Barium) | passes test |
Ca (Calcium) | ≤ 0.002 % |
K (Potassium) | ≤ 0.01 % |
Fe (Iron) | ≤ 0.0002 % |
Magnesium and alkaline-earth metals (as Ca) | ≤ 0.0100 % |
Residual solvents (ICH Q3C) | excluded by the manufacturing process |
Loss on drying (105 °C, 2 h) | ≤ 0.5 % |
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*). | |
Corresponds to Ph Eur, BP, USP |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...