Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP)

Supplier: AAT BIOQUEST INC
100411T0 100411T1 100411T2
77023-046EA 282.17 CAD
77023-046 77021-864 77022-100
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PerCP)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR