Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE/Cy7)

Supplier: AAT BIOQUEST INC
100411N0 100411N1 100411N2
77024-610EA 332.91 CAD
77024-610 77022-680 77021-464
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE/Cy7)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PE/Cy7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR