Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE/Cy5)

Supplier: AAT BIOQUEST INC
100331M0 100331M1 100331M2
77023-218EA 332.91 CAD
77023-218 77022-278 77021-522
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PE/Cy5)
Antibodies
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: PE/Cy5
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR