Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 710)

Supplier: AAT BIOQUEST INC
100330K0 100330K1
77022-812EA 385.9 CAD
77022-812 77024-976
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (iFluor® 710)
Antibodies
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: iFluor® 710
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD3e
Environmentally Preferable
Antigen name CD3e antigen
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 710
Clone SK7
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR