Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/Texas Red®)

Supplier: AAT BIOQUEST INC
100321S0 100321S1 100321S2
77022-380EA 332.91 CAD
77022-380 77023-418 77023-202
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/Texas Red®)
Antibodies
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/Texas Red®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR