Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 647)

Supplier: AAT BIOQUEST INC
100321Q0 100321Q1 100321Q2
77023-452EA 332.91 CAD
77023-452 77021-074 77024-232
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 647)
Antibodies
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/iFluor® 647
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR