Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ UV375)

Supplier: AAT BIOQUEST INC
100320X0 100320X1
77024-050EA 385.9 CAD
77024-050 77022-768
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (mFluor™ UV375)
Antibodies
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: mFluor™ UV375
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR