PYREX® Volumetric Flasks, Polyethylene Stopper, Class A, Corning
Supplier: Corning
5642-5 5642-10 5642-100 5642-200 5642-1L 5642-500 5642-250 5642-25 5642-2L 5642-50
29619-198CS 78.36 CAD
29619-198 89091-098 89090-602 89090-676 89090-710 89090-748 89090-766 89090-814 89090-842 89090-908
PYREX® Volumetric Flasks, Polyethylene Stopper, Class A, Corning
Flasks Volumetric Flasks
PYREX® borosilicate glass, clear, with LDPE stoppers.
  • Graduation line is sharp and permanent
  • White marks and inscriptions are easy to read
Volumetric flasks is made with increased strength through machine blown bodies. Heavy tubing rings and heavy beaded seal is tooled for polyethylene tubing.