Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 633)

Supplier: AAT BIOQUEST INC
100210E0 100210E1
77025-028EA 385.9 CAD
77025-028 77024-916
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 633)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 633
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD2
Environmentally Preferable
Antigen name CD2 antigen
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 633
Clone RPA-2.10
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR