Sodium hydrogen carbonate 99.0-101.0%, EMPROVE® ESSENTIAL E500, USP, JP, Ph. Eur., BP, FCC, extra pure, SAFC®

Supplier: MilliporeSigma

Synonyms: Sodium bicarbonate

1.06323.2500 1.06323.2500 1.06323.9029 1.06323.9500 1.06323.9500
CA1.06323.2500EA 1099.89 CAD
CA1.06323.2500 EM1.06323.2500 CA1.06323.9029 CA1.06323.9500 EM1.06323.9500
Sodium hydrogen carbonate 99.0-101.0%, EMPROVE® ESSENTIAL E500, USP, JP, Ph. Eur., BP, FCC, extra pure, SAFC®
Sodium hydrogen carbonate

Typical particle size distribution; 99% <75 mesh; 60% <150 mesh.

Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) 99.0-101.0%
Assay (acidimetric; calculated on dried substance) 99.5-100.5%
Aluminium (Al) 0.00005% max
Ammonium (NH4) Passes
Appearance of solution Passes
Arsenic (As) 0.0002% max
Calcium (Ca) 0.01% max
Carbonate Passes
Chloride (Cl) 0.015% max
Copper (Cu) 0.0005% max
Heavy metals (as Pb) 0.0005% max
Identification Passes
Insoluble matter Passes
Iron (Fe) 0.002% max
Lead (Pb) 0.0002% max
Loss on drying (Silica gel) 0.25% max
Mercury (Hg) 0.0001% max
pH (1%, water) 8.0-8.6
pH (5%, water) 7.9-8.4
Residual Solvents (Ph.Eur./USP/ICH) Excluded by production process
Sulfate (SO4) 0.01% max
Sulfur compounds (as SO4) 0.015% max
Zinc (Zn) 0.0025% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR