KIMBLE® RAY-SORB® Amber Class A Volumetric Flask, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
28016-10 28016-100 28016-200 28016-25 28016-250 28016-50 28016-500
89002-008CS 1448.37 CAD
89002-008 89002-010 89002-012 89002-014 89002-016 89002-018 89002-020 89002-022
KIMBLE® RAY-SORB® Amber Class A Volumetric Flask, DWK Life Sciences
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288, Class A unserialized requirements.
RAY-SORB® process protects materials sensitive to light of the shorter wavelengths. Resistant to mechanical and thermal shock. Supplied with a [ST] PTFE stopper and color-coded handle. Flasks include a white marking spot.