Potassium bromide ≥99.0%, BAKER INSTRA-ANALYZED®, J.T.Baker®

Supplier: VWR International
Warning


Management of Change (MOC) category = N

BAKR2961-03 BAKR2961-05
CAJT2961-3EA 507.81 CAD
CAJT2961-3 CAJT2961-5
Potassium bromide ≥99.0%, BAKER INSTRA-ANALYZED®, J.T.Baker®
Potassium bromide

For infrared analysis. Lot analysis on label.

Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (KBr) (by Ag titrn) ≥99.0 %
Insoluble Matter ≤0.005 %
Loss on Drying at 105°C ≤0.05 %
pH of 5% Solution at 25°C 5.0 - 7.5
Nitrogen Compounds (as N) ≤0.0005 %
Sulfate (SO₄) ≤0.002 %
Suitability for Infrared Analysis Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR