Potassium acetate ≥99% (dried basis), granules USP, Multi-Compendial endotoxin tested, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Acetic acid potassium salt

Management of Change (MOC) category = R

BAKR2917-07 BAKR2917-R BAKR2917-05 BAKR2917-01
CAJT2917-7EA 4144.04 CAD
CAJT2917-7 CAJT2917-R CAJT2917-5 CAJT2917-01
Potassium acetate ≥99% (dried basis), granules USP, Multi-Compendial endotoxin tested, J.T.Baker®
Potassium acetate

Granular. Pyrogen tested. GMP manufactured. Lot analysis provided.


  • GMP manufactured
  • Lot analysis provided.

Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (CH₃COOK) (dried basis) 99.0 - 100.5 %
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
USP - Identification Passes Test
USP - Loss on Drying ≤1.0 %
USP - pH (1 in 20) 7.5 - 8.5
USP - Residual Acetic Acid, For Information Only
USP - Sodium (Na) ≤0.03 %
EP/BP - Assay (CH₃COOK) (dried basis) 99.0 - 101.0 %
EP/BP - Aluminum (Al) ≤1 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 7.5 - 9.0
EP/BP - Reducing Substances Passes Test
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Iron (Fe) ≤20 ppm
EP/BP - Sodium (Na) ≤0.50 %
EP/BP - Loss on Drying ≤3.0 %
Additional Analytical Control - Arsenic (As) ≤8 ppm
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR