Sulfuric acid 25% (by alkalimetry), Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
1.00716.1000
1.00716.9025
CA1.00716.1000EA
73
CAD
CA1.00716.1000
EM1.00716.9025
Sulfuric acid 25% (by alkalimetry), Supelco®
Sulfuric acid
For general laboratory use.
Formula:
H₂SO₄ Boiling Pt: ∼100 °C (1013 hPa) Density: 1.1…1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Aluminum (Al) | 0.05 ppm max. |
Ammonium (NH4) | 2 ppm max. |
Arsenic (As) | 0.01 ppm max. |
Barium (Ba) | 0.05 ppm max. |
Beryllium (Be) | 0.01 ppm max. |
Bismuth (Bi) | 0.1 ppm max. |
Cadmium (Cd) | 0.02 ppm max. |
Calcium (Ca) | 0.2 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...